Learn More
Sino Biological Anti-Factor H Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10714MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Factor H (rh Complement Factor H; Catalog#10714-H08H; NP_000177.2; Ser860-Arg1231). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0653