Learn More
Sino Biological Human CFH MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10714MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Factor H / CFH (rh Complement Factor H / CFH; Catalog#10714-H08H; NP_000177.2; Ser 860-Arg 1231). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2079