Learn More
Sino Biological Anti-Factor H-related 1/CFHR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102479T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CFHR1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1065