Learn More
Sino Biological Human CFHR2 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502371856
Encompass_Preferred
Each vial contains 25 ng of recombinant Human CFHR2. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
Catalog No. 50-237-1856 Supplier Sino Biological Supplier No. SEK139205
Each vial contains 25 ng of recombinant Human CFHR2. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.