Learn More
Sino Biological Anti-Chorionic Gonadotropin/CGA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10903MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CGA / HCG (rh CGA / HCG; Catalog#10903-H08H; NP_000726.1; Met1-Ser116). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5080