Learn More
Sino Biological Human CGA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 90025C07H1
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CGA / HCG (rh CGA / HCG; Catalog#10903-H08H; NP_000726.1; Met1-Ser116). CGA / HCG specific IgG was purified by Human CGA / HCG affinity chromatography.
Catalog No. 50-237-4543