Learn More
Sino Biological Human CGA Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10903H08H50
Encompass_Preferred
A DNA sequence encoding the human CGA (NP_000726.1) (Met1-Ser116) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-237-3296