Learn More
Sino Biological Human CGREF1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13360T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CGREF1 (rh CGREF1; Catalog#13360-H08H; Q99674-1; Met1-Ile301). CGREF1 specific IgG was purified by Human CGREF1 affinity chromatography.
Catalog No. 50-237-1071