Learn More
Raybiotech Inc Human/Mouse/Rat CGRP-I EIA Kit.
Supplier:  Raybiotech Inc EIACGRP1
Encompass_Preferred
Optimized for serum, plasma and cell culture media. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8808