Learn More
Sino Biological Human CHAD PAb 100ul (Sino Biological)
Supplier:  Sino Biological 201350T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CHAD fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-1640