Learn More
Sino Biological Phospho-Chk1 (Ser345) Antibody, Rabbit MAb
Supplier:  Sino Biological 110565R000920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to residues around (Ser345) of Human Phospho-Chk1.
Catalog No. 50-241-2501