Learn More
Sino Biological Anti-Chk1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10539T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Chk1 (rh Chk1; Catalog#10539-H09B; AAM78553.1; Met1-Thr476). Chk1 specific IgG was purified by Human Chk1 affinity chromatography.
Catalog No. 50-240-8856