Learn More
Sino Biological Human CHEK1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10539T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Chk1 (rh Chk1; Catalog#10539-H09B; AAM78553.1; Met1-Thr476). Chk1 specific IgG was purified by Human Chk1 affinity chromatography.
Catalog No. 50-237-0719