Learn More
Raybiotech Inc C-Series Human Chemokine Antibody Array 1 Kit.
Catalog No. 501948524
Encompass_Preferred
Detects 38 Human Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BLC (CXCL13), CCL28 (MEC), Ck beta 8-1 (CCL23), CTACK (CCL27), CXCL16, ENA-78 (CXCL5), Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Eotaxin-3 (CCL26), Fractalkine (CX3CL1), GCP-2 (CXCL6), GRO alpha/beta/gamma, GRO alpha (CXCL1), HCC-4 (CCL16), I-309 (TCA-3/CCL1), I-TAC (CXCL11), IL-8 (CXCL8), IP-10 (CXCL10), Lymphotactin (XCL1), MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-1 delta (CCL15), MIP-3 alpha (CCL20), MIP-3 beta (CCL19), MPIF-1 (CCL23), NAP-2 (PPBP/CXCL7)
Catalog No. 50-194-8524 Supplier Raybiotech Inc Supplier No. AAHCHE12
Detects 38 Human Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BLC (CXCL13), CCL28 (MEC), Ck beta 8-1 (CCL23), CTACK (CCL27), CXCL16, ENA-78 (CXCL5), Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Eotaxin-3 (CCL26), Fractalkine (CX3CL1), GCP-2 (CXCL6), GRO alpha/beta/gamma, GRO alpha (CXCL1), HCC-4 (CCL16), I-309 (TCA-3/CCL1), I-TAC (CXCL11), IL-8 (CXCL8), IP-10 (CXCL10), Lymphotactin (XCL1), MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MDC (CCL22), MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-1 delta (CCL15), MIP-3 alpha (CCL20), MIP-3 beta (CCL19), MPIF-1 (CCL23), NAP-2 (PPBP/CXCL7)