Learn More
Raybiotech Inc G-Series Human Chemokine Array 1 Kit.
Supplier:  Raybiotech Inc GSHCHE11
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 6Ckine (CCL21), Axl, Betacellulin (BTC), CCL28 (MEC), CTACK (CCL27), CXCL16, ENA-78 (CXCL5), Eotaxin-3 (CCL26), GCP-2 (CXCL6), GRO, HCC-1 (CCL14), HCC-4 (CCL16), IL-9, IL-17F, IL-18 BP alpha, IL-28A, IL-29, IL-31, IP-10 (CXCL10), I-TAC (CXCL11), LIF, Light (TNFSF14), Lymphotactin (XCL1), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MDC (CCL22), MIF, MIP-3 alpha (CCL20), MIP-3 beta (CCL19), MPIF-1 (CCL23), MSP alpha/beta, NAP-2 (PPBP/CXCL7), Osteopontin (SPP1), PARC (CCL18), Platelet Factor 4 (CXCL4), SDF-1 alpha (CXCL12 alpha), TARC (CCL17), TECK (CCL25), TSLP
Catalog No. 50-195-0380