Learn More
American Research Products Inc Human chemokine C X C motif
Catalog No. 5000327758
Encompass
Human chemokine (C-X-C motif) ligand 17 (CXCL17) ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates urine tissue homogenates Method Sandwich
Catalog No. 50-003-27758 Supplier American Research Products Inc Supplier No. CSBEL006246HU
Human chemokine (C-X-C motif) ligand 17 (CXCL17) ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates urine tissue homogenates Method Sandwich