Learn More
Biomatik Corporation Human Chemokine C-Motif Ligand 2 (XCL2) CLIA Kit
Supplier:  Biomatik Corporation EKU09875
Encompass_Preferred
Human Chemokine C-Motif Ligand 2 (XCL2) CLIA Kit for detecting XCL2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UBD3. Gene ID: 6846. Target Synonyms: SCYC2; SCM-1b; Small Inducible Cytokine Subfamily C Member 2; Cytokine SCM-1 beta; C motif chemokine 2; XC chemokine ligand 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8412