Learn More
Biomatik Corporation Human Chemokine C-X3-C-Motif Ligand 1 (CX3CL1) CLIA Kit
Supplier:  Biomatik Corporation EKU09434
Encompass_Preferred
Human Chemokine C-X3-C-Motif Ligand 1 (CX3CL1) CLIA Kit for detecting CX3CL1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.03pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P78423. Gene ID: 6376. Target Synonyms: NTN; ABCD3; C3Xkine; CXC3; CXC3C; NTT; SCYD1; ABCD3; FKN; Neurotactin; Fractalkine; Small Inducible Cytokine Subfamily D (Cys-X3-Cys) Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5642