Learn More
Biomatik Corporation Human Chemokine (C-X-C Motif) Ligand 1 (CXCL1) CLIA Kit
Supplier:  Biomatik Corporation EKU09731
Encompass_Preferred
Human Chemokine (C-X-C Motif) Ligand 1 (CXCL1) CLIA Kit for detecting C-X-C Motif in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09341. Gene ID: 2919. Target Synonyms: NAP3; GRO1; GROa; GRO-A; MGSA-A; NAP3; SCYB1; Chemokine C-X-C-Motif Ligand 1; Melanoma Growth Stimulating Activity Alpha; Fibroblast Secretory Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5018