Learn More
Biomatik Corporation Human Chemokine (C-X-C Motif) Ligand 14 (CXCL14) ELISA Kit
Supplier:  Biomatik Corporation EKU02802
Encompass_Preferred
Human Chemokine (C-X-C Motif) Ligand 14 (CXCL14) ELISA Kit for detecting C-X-C Motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95715. Gene ID: 9547. Target Synonyms: BRAK; SCYB14; BRAK; NJAC; Bolekine; Kec; MIP2-G; MIP2g; BMAC; KS1; Breast And Kidney Expressed Chemokine; Small Inducible Cytokine Subfamily B (Cys-X-Cys) Member 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5045