Learn More
Biomatik Corporation Human Chemokine Like Factor Superfamily 6 (CKLFSF6) ELISA Kit
Supplier:  Biomatik Corporation EKU10715
Encompass_Preferred
Human Chemokine Like Factor Superfamily 6 (CKLFSF6) ELISA Kit for detecting CKLFSF6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NX76. Gene ID: 54918. Target Synonyms: CMTM6; CKLF-Like MARVEL Transmembrane Domain Containing 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8306