Learn More
BIOHIPPO Human CHGB Microsample
SDP
Catalog No. 5000315004
Encompass
Human CHGB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGB in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 244 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CHGB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CHGB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CHGB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15004 Supplier BIOHIPPO Supplier No. BHE15211776
Human CHGB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGB in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 244 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CHGB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CHGB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CHGB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color