Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CHI3L1 Recombinant Protein, PeproTech®
GREENER_CHOICE

Catalog No. 5000331933
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-003-31933 500 μg
50-003-31931 100 μg
45-065-1MG 1 mg
50-003-31932 250 μg
45-065-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-003-31933 Supplier Gibco™ Supplier No. 45065500UG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human Chitinase-3-like protein 1 (CGP39) is a 40.5 kDa monomeric protein containing 362 amino-acid resideus comprised of the mature sequence of the protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36222
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 1116
Molecular Weight (g/mol) 40.5 kDa
Name Human CHI3L1
Purification Method Purified
Quantity 500 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 38 kDa heparin-binding glycoprotein; 38kDa heparin-binding glycoprotein; 39 kDa synovial protein; 39 kDa whey protein; 40 kDa secretory signalling glycoprotein; ASRT7; AW208766; BP40; BP40 precursor; breast regression protein 39; Brp39; BRP39 protein; Cartilage glycoprotein 39; cartilage glycoprotein-39; CGP39; CGP-39; CHI3L1; Chil1; chitinase 3 like 1; Chitinase 3 like protein 1CGP 39; chitinase 3-like 1; chitinase 3-like 1 (cartilage glycoprotein-39); chitinase 3-like 1 protein; chitinase-3-like protein 1; chitinase-like 1; chitinase-like protein 1; CLP-1; DKFZp686N19119; FLJ38139; GP38K; Gp39; GP-39; hCGP39; HC-gp39; hCGP-39; HCGP-3P; Mammary gland protein 40; MGP-40; signal processing protein; signal-processing protein; SPC-40; SPS-40; YKL40; YKL-40; YYL-40
Common Name CHI3L1
Gene Symbol CHI3L1
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence YKLVCYYTSW SQYREGDGSC FPDALDRFLC THIIYSFANI SNDHIDTWEW NDVTLYGMLN TLKNRNPNLK TLLSVGGWNF GSQRFSKIAS NTQSRRTFIK SVPPFLRTHG FDGLDLAWLY PGRRDKQHFT TLIKEMKAEF IKEAQPGKKQ LLLSAALSAG KVTIDSSYDI AKISQHLDFI SIMTYDFHGA WRGTTGHHSP LFRGQEDASP DRFSNTDYAV GYMLRLGAPA SKLVMGIPTF GRSFTLASSE TGVGAPISGP GIPGRFTKEA GTLAYYEICD FLRGATVHRI LGQQVPYATK GNQWVGYDDQ ESVKSKVQYL KDRQLAGAMV WALDLDDFQG SFCGQDLRFP LTNAIKDALA AT
Activity Determined by its ability to support adhesion of Human FaDu cells.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.