Learn More
Sino Biological Anti-CHL-1/L1CAM-2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10143MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CHL1 / LICAM2 / CALL (rh CHL1 / LICAM2 / CALL; Catalog#10143-H08H; AAI04919.1; Met1-Gln1080). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8394