Learn More
Sino Biological Anti-CHL-1/L1CAM-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10143RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CHL1 (rh CHL1; Catalog#10143-H08H; AAI04919.1; Met 1-Gln 1080). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2258