Learn More
Biomatik Corporation Human Chloride Intracellular Channel Protein 4 (CLIC4) ELISA Kit
Supplier:  Biomatik Corporation EKU03130
Encompass_Preferred
Human Chloride Intracellular Channel Protein 4 (CLIC4) ELISA Kit for detecting CLIC4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y696. Gene ID: 25932. Target Synonyms: H1; CLIC4L; huH1; p64H1; Intracellular chloride ion channel protein p64H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5939