Learn More
Sino Biological Anti-CHMP4A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204868T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CHMP4A fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-0531