Learn More
Biomatik Corporation Human Cholecystokinin 18 (CCK18) ELISA Kit
Supplier:  Biomatik Corporation EKU03137
Encompass_Preferred
Human Cholecystokinin 18 (CCK18) ELISA Kit for detecting CCK18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.24pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06307. Gene ID: 885. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4436