Learn More
Biomatik Corporation Human Cholesterol-25-Hydroxylase (CH25H) ELISA Kit
Supplier:  Biomatik Corporation EKU03151
Encompass_Preferred
Human Cholesterol-25-Hydroxylase (CH25H) ELISA Kit for detecting CH25H in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95992. Gene ID: 9023. Target Synonyms: h25OH; Cholesterol 25-monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9061