Learn More
Biomatik Corporation Human Cholinergic Receptor, Muscarinic 2 (CHRM2) ELISA Kit
Supplier:  Biomatik Corporation EKU03157
Encompass_Preferred
Human Cholinergic Receptor, Muscarinic 2 (CHRM2) ELISA Kit for detecting CHRM2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08172. Gene ID: 1129. Target Synonyms: CHR-M2; HM2; M-AChRM2; Muscarinic Acetylcholine Receptor M2; Cholinergic Receptor,Muscarinic 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5890