Learn More
Biomatik Corporation Human Cholinergic Receptor, Nicotinic, Beta 3 (CHRNb3) ELISA Kit
Supplier:  Biomatik Corporation EKU03168
Encompass_Preferred
Human Cholinergic Receptor, Nicotinic, Beta 3 (CHRNb3) ELISA Kit for detecting CHRNb3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05901. Gene ID: 1142. Target Synonyms: CHR-NB3; CHRN-B3; N-AChRB3; NAChRB3; N-AChR-B3; Neuronal Acetylcholine Receptor Beta 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7192