Learn More
Biomatik Corporation Human Cholinergic Receptor, Nicotinic, Beta 2 (CHRNb2) ELISA Kit
Supplier:  Biomatik Corporation EKU03167
Encompass_Preferred
Human Cholinergic Receptor, Nicotinic, Beta 2 (CHRNb2) ELISA Kit for detecting CHRNb2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17787. Gene ID: 1141. Target Synonyms: CHR-NB2; CHRN-B2; EFNL3; NAChRB2; N-AChR-B2; N-AChRB2; Neuronal Acetylcholine Receptor Beta 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9059