Learn More
Invitrogen™ Human Chordin-Like-1/CHRDL1 ELISA Kit
Description
Human Chordin-Like-1/CHRDL1 quantitates human Chordin-Like-1/CHRDL1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Chordin-Like-1/CHRDL1.
Specifications
Specifications
| Accession Number | Q9BU40 |
| Assay Range | 32.77-8000 pg/mL |
| Assay Sensitivity | 32 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | CHL; CHL1; chordin like 1; chordin-like 1; chordin-like protein 1; Chrdl1; dA141H5.1; Kjr; Kohjirin; MGC1; MGCN; neuralin 1; neuralin-1; neurogenesin 1; Neurogenesin-1; Ng1; Nrln1; Ventroptin; ventroptin-alpha; ventroptin-beta; VOPTCHL; CHL1; chordin like 1; chordin-like 1; chordin-like protein 1; Chrdl1; dA141H5.1; Kjr; Kohjirin; MGC1; MGCN; neuralin 1; neuralin-1; neurogenesin 1; Neurogenesin-1; Ng1; Nrln1; Ventroptin; ventroptin-alpha; ventroptin-beta; VOPT; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.