Learn More
Biomatik Corporation Human Chordin Like Protein 1 (CHRDL1) ELISA Kit
Supplier:  Biomatik Corporation EKU03171
Encompass_Preferred
Human Chordin Like Protein 1 (CHRDL1) ELISA Kit for detecting CHRDL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BU40. Gene ID: 91851. Target Synonyms: NRLN1; CHL; VOPT; Ventroptin; Neuralin-1; Neurogenesin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5347