Learn More
Biomatik Corporation Human Chorionic Gonadotrophin, Hcg Elisa Kit
Supplier:  Biomatik Corporation EKC31196
Encompass_Preferred
Human Chorionic Gonadotrophin, Hcg Elisa Kit for detecting Chorionic Gonadotrophin, Hcg in serum, plasma, urine, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 8-240mIU/ml. Sensitivity: 2mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P0DN86; CGB3. Target Synonyms: CGB3; CGB;; CGB5;; CGB8; Choriogonadotropin subunit beta 3; Choriogonadotropin subunit beta; CG-beta; Chorionic gonadotropin chain beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5561