Learn More
Biomatik Corporation Human Chorionic Gonadotropin Alpha Polypeptide (CGa) ELISA Kit
Supplier:  Biomatik Corporation EKU08537
Encompass_Preferred
Human Chorionic Gonadotropin Alpha Polypeptide (CGa) ELISA Kit for detecting CGa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.5-10,000pg/ml. Sensitivity: 50.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01215. Gene ID: 1081. Target Synonyms: FSHA; GPHA1; GPHa; HCG; LHA; TSHA; Thyroid Stimulating Hormone Alpha; Follicle-Stimulating Hormone Alpha Subunit; Luteinizing hormone alpha chain; Lutropin alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8657