Learn More
Biomatik Corporation Human Chorionic Gonadotropin Beta Polypeptide (CGb) ELISA Kit
Supplier:  Biomatik Corporation EKU08747
Encompass_Preferred
Human Chorionic Gonadotropin Beta Polypeptide (CGb) ELISA Kit for detecting CGb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1000ng/ml. Sensitivity: 4.64ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P0DN86; P01233. Gene ID: 1082; 1082. Target Synonyms: CG-B; CGB3; hCGB; HCG; CGb5; CGb7; CGb8; Choriogonadotropin subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7042