Learn More
BIOHIPPO Human CHRD Microsample
SDP
Catalog No. 5000315018
Encompass
Human CHRD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHRD in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CHRD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CHRD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CHRD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15018 Supplier BIOHIPPO Supplier No. BHE15211285
Human CHRD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHRD in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CHRD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CHRD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CHRD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color