Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CHRND (aa 374-465) Control Fragment Recombinant Protein

Catalog No. RP108416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108416 Supplier Invitrogen™ Supplier No. RP108416
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The acetylcholine receptor of muscle has 5 subunits of 4 different types: 2 alpha and 1 each of beta, gamma and delta subunits. After acetylcholine binding, the receptor undergoes an extensive conformation change that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07001
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 1144
Name Human CHRND (aa 374-465) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias acetylcholine receptor; acetylcholine receptor delta subunit; acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); AChR delta subunit; Achr-4; ACHRD; Acrd; cholinergic receptor; cholinergic receptor nicotinic delta subunit; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; delta subunit of muscle acetylcholine receptor; FCCMS; L10076; nicotinic acetylcholine receptor delta polypeptide; SCCMS; sofa potato; sop; wu:fa01e01; zgc:110469
Common Name CHRND
Gene Symbol CHRND
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LVRRSSSLGYISKAEEYFLLKSRSDLMFEKQSERHGLARRLTTARRPPASSEQAQQELFNELKPAVDGANFIVNHMRDQNNYNEEKDSWNRV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.