Learn More
Biomatik Corporation Human Chromobox Homolog 3 (CBX3) ELISA Kit
Supplier:  Biomatik Corporation EKU03177
Encompass_Preferred
Human Chromobox Homolog 3 (CBX3) ELISA Kit for detecting CBX3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13185. Gene ID: 11335. Target Synonyms: HECH; HP1-GAMMA; HP1Hs-Gamma; HP1 Gamma Homolog,Drosophila; Heterochromatin protein 1 homolog gamma; Modifier 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7183