Learn More
Biomatik Corporation Human Chymotrypsinogen B2 (CTRB2) ELISA Kit
Supplier:  Biomatik Corporation EKU03189
Encompass_Preferred
Human Chymotrypsinogen B2 (CTRB2) ELISA Kit for detecting CTRB2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6GPI1. Gene ID: 440387. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7190