Learn More
Raybiotech Inc Human cIAP-2 ELISA Kit
Supplier:  Raybiotech Inc ELHCIAP21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human CIAP-2. Other species not determined yet.
Catalog No. 50-194-9123