Learn More
Biomatik Corporation Human Ciliary Neurotrophic Factor Receptor (CNTFR) ELISA Kit
Supplier:  Biomatik Corporation EKU11709
Encompass_Preferred
Human Ciliary Neurotrophic Factor Receptor (CNTFR) ELISA Kit for detecting CNTFR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26992. Gene ID: 1271. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6968