Learn More
Biomatik Corporation Human Citrate Synthase (CS) ELISA Kit
Supplier:  Biomatik Corporation EKU03196
Encompass_Preferred
Human Citrate Synthase (CS) ELISA Kit for detecting CS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75390. Gene ID: 1431. Target Synonyms: Citrate (Si)-synthase, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5164