Learn More
BIOHIPPO Human CK-18 Microsample
SDP
Catalog No. 5000314342
Encompass
Human CK-18 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CK-18 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CK-18 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CK-18 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CK-18 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14342 Supplier BIOHIPPO Supplier No. BHE15210678
Human CK-18 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CK-18 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CK-18 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CK-18 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CK-18 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color