Learn More
BIOHIPPO Human CK Microsample
SDP
Catalog No. 5000314106
Encompass
Human CK Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CK in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 IU/mL with a sensitivity of 7 IU/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CK 18-M30 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CK 18-M30 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CK 18-M30 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14106 Supplier BIOHIPPO Supplier No. BHE15212363
Human CK Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CK in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 IU/mL with a sensitivity of 7 IU/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CK 18-M30 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CK 18-M30 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CK 18-M30 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color