Learn More
Sino Biological Human CKMT1A PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13924RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CKMT1A (rh CKMT1A; Catalog#13924-H07B1; P12532-1; Ala40-His417). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2298