Learn More
ACROBiosystems Human Claudin-1 100ug
Catalog No. 502535258
Encompass_Preferred
Human Claudin-1 Full Length Protein (VLP) (CL1-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 211 (Accession # O95832).
Catalog No. 50-253-5258 Supplier ACROBiosystems Supplier No. CL1H52P6100UG
Human Claudin-1 Full Length Protein (VLP) (CL1-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 211 (Accession # O95832).