Learn More
ACROBiosystems Human Claudin-1 500ug
Catalog No. 502535253
Encompass_Preferred
Human Claudin-1 Protein FlagHis Tag (CL1-H55D8) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Val 211 (Accession # O95832).
Catalog No. 50-253-5253 Supplier ACROBiosystems Supplier No. CL1H55D8500UG
Add to Cart
Add to Cart
Request a Quote
Human Claudin-1 Protein FlagHis Tag (CL1-H55D8) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Val 211 (Accession # O95832).