Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human/Cynomolgus Claudin-18.2 Protein, His and Twin-Strep Tag (Nanodisc), 100 μg

Catalog No. 502370252
Encompass_Preferred

Recombinant human/cynomolgus Claudin-18.2 in a nanodisc format for stabilized study of this integral membrane protein. Expressed in baculovirus-insect cells with N-terminal His and C-terminal twin-Strep tags; supplied as a 100 μg vial in buffered solution.

  • Nanodisc format for membrane protein stability.
  • Expression system: baculovirus-insect cells.
  • Tags: N-terminal His tag and C-terminal twin-Strep tag.
  • Amino acids Met 1-Ala 200; calculated molecular weight ~26.5 kDa.
  • Supplied as 100 μg in 20 mM HEPES, 150 mM NaCl, pH 7.5, with trehalose; 0.2 μm filtered.
  • Purity greater than 80% by SDS-PAGE; endotoxin below 1.0 EU/μg.
  • Validated for ELISA, SPR, and flow cytometry (QC tested).

Catalog No. 50-237-0252 Supplier ACROBiosystems Supplier No. CL2H5586100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

Recombinant human/cynomolgus Claudin-18.2 in a nanodisc format for stabilized study of this integral membrane protein. Expressed in baculovirus-insect cells with N-terminal His and C-terminal twin-Strep tags; supplied as a 100 μg vial in buffered solution.

  • Nanodisc format for membrane protein stability.
  • Expression system: baculovirus-insect cells.
  • Tags: N-terminal His tag and C-terminal twin-Strep tag.
  • Amino acids Met 1-Ala 200; calculated molecular weight ~26.5 kDa.
  • Supplied as 100 μg in 20 mM HEPES, 150 mM NaCl, pH 7.5, with trehalose; 0.2 μm filtered.
  • Purity greater than 80% by SDS-PAGE; endotoxin below 1.0 EU/μg.
  • Validated for ELISA, SPR, and flow cytometry (QC tested).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.